missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Canine IL-31 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
0.1 mg
250 μg
Unit Size:
0.05mg
0.1mg
250µg
Description
A recombinant protein with a un-tagged and corresponding to the amino acids 24-159aa of Canine IL31.
Source: Baculovirus
Amino Acid Sequence:
SHMAPTHQLP PSDVRKIILE LQPLSRGLLE DYQKKETGVP ESNRTLLLCL TSDSQPPRLN SSAILPYFRA IRPLSDKNII DKIIEQLDKL KFQHEPETEI SVPADTFECK SFILTILQQF SACLESVFKS LNSGPQ
Spezifikation
Spezifikation
| Concentration | 0.25 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | PBS (pH 7.4), 20% glycerol |
| Gene ID (Entrez) | 386653 |
| Molecular Weight (g/mol) | 15.3 kDa |
| Quantity | 0.05 mg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | IL-31interleukin-31, interleukin 31 |
| Gene Symbol | IL31 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu