missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken GH Protein

Product Code. 15984468
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
This item is not returnable. View return policy

Product Code. 15984468

Brand: enQuireBio™ QP106321mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.

Specifications

Name GH Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity GH Chicken Recombinantis fully biologically active in homologous assays and in PDF-P1 3B9 cells stably transfected with rabbit GH receptors.
Product Type Recombinant Protein
Cross Reactivity Chicken
Species E. coli
Protein Tag Untagged
Sequence MAPGSWFSPL LIAVVTLGLP QEAAATFPAM PLSNLFANAV LRAQHLHLLA AETYKEFERT YIPEDQRYTN KNSQAAFCYS ETIPAPTGKD DAQQKSDMEL LRFSLVLIQS WLTPVQYLSK VFTNNLVFGT SDRVFEKLKD LEEGIQALMR ELEDRSPRGP QLLRPTYDKF DIHLRNEDAL LKNYGLLSCF KKDLHKVETY LKVMKCRRFG ESNCTI
Buffer The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.3% NaHCO3 adjusted to pH 8.
Purity or Quality Grade Greater than 99.0% as determined by:(a) Analysis by SDS-PAGE gel. (b) Analysis by SEC-HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.