missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Vitellogenin-1 Protein

Product Code. 16056940
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16056940 50 μg 50µg
16016940 10 μg 10µg
16006940 100 μg 100µg
16036940 200 μg 200µg
16046940 500 μg 500µg
16026940 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16056940 Supplier enQuireBio™ Supplier No. QP699150ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Gallus gallus (Chicken) Vitellogenin-1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001004408.1
Gene ID (Entrez) 424547
Name Vitellogenin-1 Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol VTG1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Chicken
Species E. coli
Protein Tag His
Sequence FGENKVYTYN YESILFSGIP EKGLARTGIR IRSEVEISGI GPKLCLIRIH SIEAAEYNGI WPTSSFSRSL KLTQALTGQL SIPIKFEYSN GHVGNLMAPD SVSDDGLNIY RGILNILELS LKKMQHSYSI QEAGIGGICN TTYAIQENKK ANLVDVTKSK DLNSCEEKVQ VVTGSAYTQP CQTCQQRNKN SRATATYNYK IKYTHNEAVI TQAEVEEVHQ FTPFHEITGG NAIVEARQKL ALIEVQKQVA EVPPKEFQKR GSLQYQFGSE LLQLPVHLFK IKDVERQIEE RLQDLVETTY EQLPSDAPAK ALKLMHLLRA ANEENYESVW KQFSSRPAYR RYLLDLLPAA ASHRSLRFLR HKMERQELTN WEIAQTVLVA LHSSSPTQEV MEEATLIVKK HCPRSSSVLR KVCLLSYASL CHKRCSSPYS CSECLQVFHV FAGEALGKSN IEEVLLALKA LGNVGHPASI KHIKKFLPGY AAGASELPLK VHETAVMALK SIGMRDPQMV QAITLEIFLN HKIHPRIRML AAVVLLETKP GLPILMILVD AVLKEPSMQV ASFIYSHLRA LGRSTAPDLQ MMASACRMAV RALSPKFDRS GYQFSKVFRF
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.