Learn More
enQuireBio™ Recombinant Chikungunya Virus E1 Envelope Protein
Recombinant Protein
Brand: enQuireBio™ QP10474-100ug
Additional Details : Weight : 0.01000kg
Specifications
E1 Envelope Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
PBS, pH7.4. |
100 μg | |
Untested | |
Chikungunya virus (Strain: Senegal/37997) | |
Native | |
>90% as determined by SDS-PAGE. |