missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chimpanzee LTB / TNFSF3 / Lymphotoxin beta Protein
A DNA sequence encoding the Pan troglodytes (Chimpanzee) LTB / TNFSF3 / Lymphotoxin beta, was expressed in the hosts and tags indicated.
€ 309.00 - € 3405.00
Specifications
Accession Number | NP_001070252.1 |
---|---|
Gene ID (Entrez) | 744486 |
Name | LTB / TNFSF3 / Lymphotoxin beta Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030801
|
enQuireBio™
QP7788-EC-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050801
|
enQuireBio™
QP7788-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040801
|
enQuireBio™
QP7788-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060801
|
enQuireBio™
QP7788-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070801
|
enQuireBio™
QP7788-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080801
|
enQuireBio™
QP7788-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001070252.1 | |
LTB / TNFSF3 / Lymphotoxin beta Protein | |
Not determined. | |
Extracellular Domain | |
Chimpanzee | |
His | |
Tris-based buffer, 50% glycerol |
744486 | |
Research Use Only | |
LTB | |
Recombinant Protein | |
E. coli | |
QDQGGLVTET ADPGAQAQQG LGFQKLPEEE PETDLSPGLP AAHLIGAPLK GQGLGWETTK EQAFLTSGTQ FSDAEGLALP QDGLYYLYCL VGYRGRTPPG GGDPQGRSVT LRSSLYRAGG AYGPGTPELL LEGAETVTPV LDPARRQGYG PLWYTSVGFG GLVQLRRGER VYVNISHPDM VDFARGKTFF GAVMVG | |
Greater than 90% as determined by SDS-PAGE. |