missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia W5-W6 Protein
A cDNA sequence encoding the Chlamydia W5-W6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11419-500ug
Additional Details : Weight : 0.01000kg
Specifications
W5-W6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-HCl, pH 7.2, 50% glycerol and 1.5M urea (1 mg/1ml). |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
Untagged | |
Protein is >90% pure as determined by SDS-PAGE. |