Learn More
R&D Systems™ Recombinant Cynomolgus alpha-L-Iduronidase/IDUA Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to cleave a fluorogenic substrate, 4-Methylumbelliferyl alpha-L-iduronide. The specific activity is >15,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | XP_005554313.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate, NaCl and Glycerol. |
| Molecular Weight (g/mol) | M.W. Observed: 81-90 kDa, under reducing conditions, M.W. Theoretical: 72 kDa |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey alpha-L-Iduronidase/IDUA protein Ala26-Pro653 with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |