Learn More
R&D Systems™ Recombinant Cynomolgus Angiopoietin-like 3 Protein
Product Code. 18758656
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18758656 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18758656 Supplier R&D Systems™ Supplier No. 10052AN050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Recombinant Cynomolgus Angiopoietin-like 3 Protein is derived from T. ni (baculovirus)
  • ANGPTL3 is a secreted glycoprotein that is structurally related to the Angiopoietins
  • Mature ANGPTL3 contains an N-terminal coiled-coil domain and a C-terminal fibrinogen-like domain.
  • ANGPTL3 is expressed in the liver from early in development through adulthood.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number XP_005543242
For Use With (Application) Bioactivity
Gene ID (Entrez) 27329
Molecular Weight (g/mol) 53 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source Trichoplusia ni, T. ni (baculovirus)-derived cynomolgus monkey Angiopoietin-like Protein 3/ANGPTL3 protein Ser17-Glu460, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water
Buffer Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and CHAPS with Trehalose
Product Line R&D Systems