missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus B7-H7/HHLA2 Fc Chimera Protein

Product Code. 18789366
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18789366 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18789366 Supplier R&D Systems™ Supplier No. 10108B7050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 secretion in human T lymphocytes. The ED50 for this effect is 1-6 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number XP_005548285
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Citric Acid and NaCl with Trehalose.
Gene ID (Entrez) 11148
Molecular Weight (g/mol) M.W. (Observed): 92-109 kDa, reducing conditions; M.W. (theoretical): 64 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey B7-H7/HHLA2 protein Cynomolgus Monkey B7-H7 (Ile21-Asn345) (N-terminus) Accession # XP_005548285 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Specific Reactivity B7-H7/HHLA2
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL aq. soln..
Buffer Lyophilized from a 0.2 μm filtered solution in Citric Acid and NaCl with Trehalose.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein B7-H7/HHLA2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.