missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus CD200R1 Fc Chimera Protein

Product Code. 18778197
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18778197

Brand: R&D Systems™ 9357CD100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Cynomolgus CD200 R1 Fc Chimera Protein is derived from HEK293. The Recombinant Cynomolgus CD200 R1 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number XP_005548208
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 131450
Molecular Weight (g/mol) 53 kDa
Name CD200R1
Quantity 100 μg
Source Human embryonic kidney cell,HEK293-derived cynomolgus monkey CD200 R1 protein, (N-terminus) Human Cynomolgus Monkey CD200 R1 (Ala27-Leu267) Accession &Num; XP_005548208 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias CD200 R1, CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, MOX2R, OX2RCD200 cell surface glycoprotein receptor
Biological Activity 1.5 to 9ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.