missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey CD40/TNFRSF5 Fc Chimera

Product Code. 18756648
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18756648

Brand: R&D Systems™ 9660CD100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey CD40/TNFRSF5 Fc Chimera is coated at 1 μg/mL, 100 μL/well, it binds to Recombinant Human CD40 Ligand/TNFSF5 (Catalog # 6420-CL ) with an ED50 of 5-25 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number XP_005569274.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 958
Molecular Weight (g/mol) M.W. (Observed): 57-64 kDa, reducing conditions; M.W. (theoretical): 46 kDa
Name B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey CD40/TNFRSF5 protein Cynomolgus Monkey CD40/TNFRSF5 (Glu21-Arg193) (N-terminus) Accession # XP_005569274.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Species Cynomolgus Monkey
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD40/TNFRSF5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.