missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cytomegalovirus Envelope glycoprotein 1
A DNA sequence encoding the Human cytomegalovirus Envelope glycoprotein L, was expressed in the hosts and tags indicated.
€ 362.00 - € 3175.00
Specifications
Accession Number | YP_081555.1 |
---|---|
Gene ID (Entrez) | 3077416 |
Name | Envelope glycoprotein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088900
|
enQuireBio™
QP7456-YE-100UG |
100 μg |
€ 1150.00
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028910
|
enQuireBio™
QP7456-YE-500UG |
500 μg |
€ 2650.00
500µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098900
|
enQuireBio™
QP7456-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018910
|
enQuireBio™
QP7456-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038910
|
enQuireBio™
QP7456-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008910
|
enQuireBio™
QP7456-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
YP_081555.1 | |
Envelope glycoprotein 1 | |
Not determined. | |
Full Length | |
Cytomegalovirus | |
His | |
Tris-based buffer, 50% glycerol |
3077416 | |
Research Use Only | |
gL | |
Recombinant Protein | |
Yeast | |
AAVSVAPTAA EKVPAECPEL TRRCLLGEVF EGDKYESWLR PLVNVTGRDG PLSQLIRYRP VTPEAANSVL LDEAFLDTLA LLYNNPDQLR ALLTLLSSDT APRWMTVMRG YSECGDGSPA VYTCVDDLCR GYDLTRLSYG RSIFTEHVLG FELVPPSLFN VVVAIRNEAT RTNRAVRLPV STAAAPEGIT LFYGLYNAVK EFCLRHQLDP PLLRHLDKYY AGLPPELKQT RVNLPAHSRY GPQAVDAR | |
Greater than 90% as determined by SDS-PAGE. |