Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 1 NS1 Protein
Recombinant Protein
Brand:  enQuireBio™ QP10475-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15943988
  • € 402.00 / 100µg
Estimated Shipment: 28-06-2024
to see stock.
Description
Description
The NS1, non-structural protein, is an immunogenic protein, but poses no health risks and is therefore ideal for antibody production.
Specifications
Specifications
Serotype 1 NS1 Protein
Research Use Only
Recombinant Protein
Human
DPBS pH 7.4.
100 μg
Untested
Dengue virus (Strain: Nauru/Western Pacific/1974)
Native
>95% as determined by SDS-PAGE.