Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 2 VLP Protein
Recombinant Protein
Brand:  enQuireBio™ QP10479-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15943998
  • € 4670.00 / 500µg
Estimated Shipment: 24-06-2024
to see stock.
Description
Description
This VLP protein is combinantion of the C, E1 and E2 sequences. Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.
Specifications
Specifications
Serotype 2 VLP Protein
Research Use Only
Recombinant Protein
Human
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4.
500 μg
Untested
Dengue virus (Strain: Thailand/16681/1984)
Native
>95% as determined by SDS-PAGE.