missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Arginine esterase Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Arginine esterase, was expressed in the hosts and tags indicated.
€ 309.00 - € 3405.00
Specifications
Accession Number | NP_001003284.1 |
---|---|
Gene ID (Entrez) | 403967 |
Name | Arginine esterase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008560
|
enQuireBio™
QP7377-EC-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028560
|
enQuireBio™
QP7377-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018560
|
enQuireBio™
QP7377-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038560
|
enQuireBio™
QP7377-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048560
|
enQuireBio™
QP7377-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058560
|
enQuireBio™
QP7377-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001003284.1 | |
Arginine esterase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
403967 | |
Research Use Only | |
KLK2 | |
Recombinant Protein | |
E. coli | |
IIGGRECLKN SQPWQVAVYH NGEFACGGVL VNPEWVLTAA HCANSNCEVW LGRHNLSESE DEGQLVQVRK SFIHPLYKTK VPRAVIRPGE DRSHDLMLLH LEEPAKITKA VRVMDLPKKE PPLGSTCYVS GWGSTDPETI FHPGSLQCVD LKLLSNNQCA KVYTQKVTKF MLCAGVLEGK KDTCKGDSGG PLICDGELVG ITSWGATPCG KPQMPSLYTR VMPHLMWIKD TMKANT | |
Greater than 90% as determined by SDS-PAGE. |