missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Epididymal secretory protein E1 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Epididymal secretory protein E1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7631-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Epididymal secretory protein E1 Protein
Research Use Only
NPC
Recombinant Protein
Yeast
VHFKDCGSAV GVIKELNVNP CPAQPCKLHK GQSYSVNVTF TSNIPSQSSK AVVHGIVLGV AVPFPIPEAD GCKSGINCPI QKDKTYSYLN KLPVKNEYPS IKLVVQWMLL GDNNQHLFCW EIPVQIEG
Greater than 90% as determined by SDS-PAGE.
50 μg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers