missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog IL4 / Interleukin-4 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.
€ 309.00 - € 3405.00
Specifications
Accession Number | NP_001003159.1 |
---|---|
Gene ID (Entrez) | 403785 |
Name | IL4 / Interleukin-4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011700
|
enQuireBio™
QP6222-EC-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031700
|
enQuireBio™
QP6222-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021700
|
enQuireBio™
QP6222-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041700
|
enQuireBio™
QP6222-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051700
|
enQuireBio™
QP6222-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061700
|
enQuireBio™
QP6222-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001003159.1 | |
IL4 / Interleukin-4 Protein | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |
403785 | |
Research Use Only | |
IL4 | |
Recombinant Protein | |
E. coli | |
HNFNITIKEI IKMLNILTAR NDSCMELTVK DVFTAPKNTS DKEIFCRAAT VLRQIYTHNC SNRYLRGLYR NLSSMANKTC SMNEIKKSTL KDFLERLKVI MQKKYYRH | |
Greater than 90% as determined by SDS-PAGE. |