missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Serum Albumin / HSA / ALB Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Serum Albumin / HSA / ALB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5642-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003026.1 | |
Serum Albumin / HSA / ALB Protein | |
Research Use Only | |
ALB | |
Recombinant Protein | |
Yeast | |
EAYKSEIAHR YNDLGEEHFR GLVLVAFSQY LQQCPFEDHV KLAKEVTEFA KACAAEESGA NCDKSLHTLF GDKLCTVASL RDKYGDMADC CEKQEPDRNE CFLAHKDDNP GFPPLVAPEP DALCAAFQDN EQLFLGKYLY EIARRHPYFY APELLYYAQQ YKGVFAECCQ AADKAACLGP KIEALREKVL LSSAKERFKC ASLQKFGDRA FKAWSVARLS QRFPKADFAE ISKVVTDLTK VHKECCHGDL LECADDRADL AKYMCENQDS ISTKLKECCD KPVLEKSQCL AEVERDELPG DLPSLAADFV EDKEVCKNYQ EAKDVFLGTF LYEYARRHPE YSVSLLLRLA KEYEATLEKC CATDDPPTCY AKVLDEFKPL VDEPQNLVKT NCELFEKLGE YGFQNALLVR YTKKAPQVST PTLVEVSRKL GKVGTKCCKK PESERMSCAE DFLSVVLNRL CVLHEKTPVS ERVTKCCSES LVNRRPCFSG LEVDETYVPK EFNAETFTFH ADLCTLPEAE KQVKKQTALV ELLKHKPKAT DEQLKTVMGD FGAFVEKCCA AENKEGCFSE EGPKLVAAAQ AALV | |
Greater than 90% as determined by SDS-PAGE. |
403550 | |
100 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |