missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Hygromycin-B 4-O-kinase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Hygromycin-B 4-O-kinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7397-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028670

  • € 2650.00 / 500µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_000742814.1
Hygromycin-B 4-O-kinase Protein
Research Use Only
hph
Recombinant Protein
Yeast
MKKPELTATS VEKFLIEKFD SVSDLMQLSE GEESRAFSFD VGGRGYVLRV NSCADGFYKD RYVYRHFASA ALPIPEVLDI GEFSESLTYC ISRRAQGVTL QDLPETELPA VLQPVAEAMD AIAAADLSQT SGFGPFGPQG IGQYTTWRDF ICAIADPHVY HWQTVMDDTV SASVAQALDE LMLWAEDCPE VRHLVHADFG SNNVLTDNGR ITAVIDWSEA MFGDSQYEVA NIFFWRPWLA CMEQQTRYFE RRHPELAGSP RLRAYMLRIG LDQLYQSLVD GNFDDAAWAQ GRCDAIVRSG AGTVGRTQIA RRSAAVWTDG CVEVLADSGN RRPSTRPRAK E
Greater than 90% as determined by SDS-PAGE.
13905269
500 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers