missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli L-lactate dehydrogenase Protein

A DNA sequence encoding the Escherichia coli O9:H4 (strain HS) L-lactate dehydrogenase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7434-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028860

  • € 3405.00 / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_000586962.1
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
L-lactate dehydrogenase Protein
Research Use Only
lldD
Recombinant Protein
E. coli
MIISAASDYR AAAQRILPPF LFHYMDGGAY SEYTLRRNVE DLSEVALRQR ILKNMSDLSL ETTLFNEKLS MPVALAPVGL CGMYARRGEV QAAKAADAHG IPFTLSTVSV CPIEEVAPAI KRPMWFQLYV LRDRGFMRNA LERAKAAGCS TLVFTVDMPT PGARYRDAHS GMSGPNAAMR RYLQAVTHPQ WAWDVGLNGR PHDLGNISAY LGKPTGLEDY IGWLGNNFDP SISWKDLEWI RDFWDGPMVI KGILDPEDAR DAVRFGADGI VVSNHGGRQL DGVLSSARAL PAIADAVKGD IAILADSGIR NGLDVVRMIA LGADTVLLGR AFLYALATAG QAGVANLLNL IEKEMKVAMT LTGAKSISEI TQDSLVQGLG KELPAALAPM AKGNAA
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers