missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Lactose operon repressor Protein
A DNA sequence encoding the Escherichia coli (strain K12) Lactose operon repressor, was expressed in the hosts and tags indicated.
€ 309.00 - € 3405.00
Specifications
Accession Number | NP_414879.3 |
---|---|
Gene ID (Entrez) | 945007 |
Name | Lactose operon repressor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008710
|
enQuireBio™
QP7406-EC-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028710
|
enQuireBio™
QP7406-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018710
|
enQuireBio™
QP7406-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038710
|
enQuireBio™
QP7406-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048710
|
enQuireBio™
QP7406-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058710
|
enQuireBio™
QP7406-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_414879.3 | |
Lactose operon repressor Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
945007 | |
Research Use Only | |
lacI | |
Recombinant Protein | |
E. coli | |
MKPVTLYDVA EYAGVSYQTV SRVVNQASHV SAKTREKVEA AMAELNYIPN RVAQQLAGKQ SLLIGVATSS LALHAPSQIV AAIKSRADQL GASVVVSMVE RSGVEACKAA VHNLLAQRVS GLIINYPLDD QDAIAVEAAC TNVPALFLDV SDQTPINSII FSHEDGTRLG VEHLVALGHQ QIALLAGPLS SVSARLRLAG WHKYLTRNQI QPIAEREGDW SAMSGFQQTM QMLNEGIVPT AMLVANDQMA LGAMRAITES GLRVGADISV VGYDDTEDSS CYIPPLTTIK QDFRLLGQTS VDRLLQLSQG QAVKGNQLLP VSLVKRKTTL APNTQTASPR ALADSLMQLA RQVSRLESGQ | |
Greater than 90% as determined by SDS-PAGE. |