missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli LexA repressor Protein

A DNA sequence encoding the Escherichia coli (strain K12) LexA repressor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7390-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028620

  • € 2645.00 / 1mg


Estimated Shipment
03-11-2020
Add to basket

Specifications

Specifications

NP_418467.1
LexA repressor Protein
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
948544
Greater than 90% as determined by SDS-PAGE.
Research Use Only
lexA
Recombinant Protein
E. coli
MKALTARQQE VFDLIRDHIS QTGMPPTRAE IAQRLGFRSP NAAEEHLKAL ARKGVIEIVS GASRGIRLLQ EEEEGLPLVG RVAAGEPLLA QQHIEGHYQV DPSLFKPNAD FLLRVSGMSM KDIGIMDGDL LAVHKTQDVR NGQVVVARID DEVTVKRLKK QGNKVELLPE NSEFKPIVVD LRQQSFTIEG LAVGVIRNGD WL
Videos
SDS
Documents