missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Methyl-accepting chemotaxis protein 1

A DNA sequence encoding the Escherichia coli (strain K12) Methyl-accepting chemotaxis protein I, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9632-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028541

  • € 2650.00 / 500µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_418775.1
Methyl-accepting chemotaxis protein 1
Research Use Only
tsr
Recombinant Protein
Yeast
WFGIKASLVA PMNRLIDSIR HIAGGDLVKP IEVDGSNEMG QLAESLRHMQ GELMRTVGDV RNGANAIYSG ASEIATGNND LSSRTEQQAA SLEETAASME QLTATVKQNA ENARQASHLA LSASETAQRG GKVVDNVVQT MRDISTSSQK IADIISVIDG IAFQTNILAL NAAVEAARAG EQGRGFAVVA GEVRNLAQRS AQAAREIKSL IEDSVGKVDV GSTLVESAGE TMAEIVSAVT RVTDIMGEIA SASDEQSRGI DQVGLAVAEM DRVTQQNAAL VEESAAAAAA LEEQASRLTE AVAVFRIQQQ QRETSAVVKT VTPAAPRKMA VADSEENWET F
Greater than 90% as determined by SDS-PAGE.
948884
500 μg
Not determined.
Cytoplasmic Domain
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers