missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ruvC Protein
A DNA sequence encoding the Escherichia coli (strain K12) ruvC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7393-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416377.1 | |
ruvC Protein | |
Research Use Only | |
ruvC | |
Recombinant Protein | |
E. coli | |
AIILGIDPGS RVTGYGVIRQ VGRQLSYLGS GCIRTKVDDL PSRLKLIYAG VTEIITQFQP DYFAIEQVFM AKNADSALKL GQARGVAIVA AVNQELPVFE YAARQVKQTV VGIGSAEKSQ VQHMVRTLLK LPANPQADAA DALAIAITHC HVSQNAMQMS ESRLNLARGR LR | |
Greater than 90% as determined by SDS-PAGE. |
946378 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |