missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli tetR Protein

A DNA sequence encoding the Escherichia coli tetR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7416-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058770

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_000088605.1
tetR Protein
Research Use Only
tetR
Recombinant Protein
E. coli
MSRLDKSKVI NSALELLNEV GIEGLTTRKL AQKLGVEQPT LYWHVKNKRA LLDALAIEML DRHHTHFCPL EGESWQDFLR NNAKSFRCAL LSHRDGAKVH LGTRPTEKQY ETLENQLAFL CQQGFSLENA LYALSAVGHF TLGCVLEDQE HQVAKEERET PTTDSMPPLL RQAIELFDHQ GAEPAFLFGL ELIICGLEKQ LKCESGS
Greater than 90% as determined by SDS-PAGE.
31515962
50 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers