missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ebola Virus Ebola Zaire Protein

Product Code. 15998158
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15998158 100 μg 100µg
15918168 500 μg 500µg
15908168 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15998158 Supplier enQuireBio™ Supplier No. QP11728100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Ebola Zaire Protein was constructed and used to recombinantly synthesize the protein.

Specifications

Name Ebola Zaire Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Ebola Virus
Species E. coli
Protein Tag His
Buffer Ebola Zaire protein solution is supplied in Phosphate buffer and 0.02% sodium azide.
Purity or Quality Grade >95% pure as determined by 12% SDS-PAGE (Coomassie blue stain).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.