missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Equine IFN g Protein

Product Code. 15934618
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15934618

Brand: enQuireBio™ QP1067520ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IFN g was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 100034181
Name IFN g Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol IFNG
Biological Activity The ED50 as determined by an anti-viral assay using human HeLa cells infected with encephalomyocarditis (EMC) virus is less than 10.0 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg.
Product Type Recombinant Protein
Cross Reactivity Equine
Species E. coli
Protein Tag Untagged
Sequence QAAFFKEIEN LKEYFNASNP DVGDGGPLFL DILKNWKEDS DKKIIQSQIV SFYFKLFENL KDNQVIQKSM DTIKEDLFVK FFNSSTSKLE DFQKLIQIPV NDLKVQRKAI SELIKVMNDL SPKANLRKRK RSQNPFRGRR ALQ
Buffer Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4 and 5% trehalose.
Purity or Quality Grade Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.