missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Equine Leptin Protein
A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 1450.00
Specifications
Name | Leptin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935068
|
enQuireBio™
QP10753-20UG |
20 μg |
€ 137.00
20µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915068
|
enQuireBio™
QP10753-100UG |
100 μg |
€ 212.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925068
|
enQuireBio™
QP10753-1MG |
1 mg |
€ 1450.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Leptin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
LWLWPYLFFI EAVPIRKVQD DTKTLIKTIV TRINDISHTQ SVSSKQRVTG LDFIPGLHPV LSLSKMDQTL AIYQQILTSL PSRNVIQISN DLENLRDLLH LLASSKSCPL PQARGLETLA SLGGVLEASL LLHRGGSPEQ AAGVS | |
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. | |
Equine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |