missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Equine Serum Amyloid A1 Protein

Product Code. 18643567 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
This item is not returnable. View return policy

Product Code. 18643567

Brand: Novus Biologicals™ NBP262533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Formulation 0.01 M HCl
Gene ID (Entrez) 6288
Molecular Weight (g/mol) MolecularWeight-theroretical: 13.6 kDa
Quantity 0.1 mg
Storage Requirements Store at -20°C.
Gene Alias MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4
Gene Symbol SAA1
Reconstitution Reconstitute with 1.0 mL distilled water. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. Gently mix vial after reconstitution.)
Research Category Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience
Purity or Quality Grade >95%
Protein Serum Amyloid A1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.