missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Gilthead Seabream GH Protein

Product Code. 15904478
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15904478

Brand: enQuireBio™ QP1063310ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.

Specifications

Name GH Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Binding assays of the 125I-labeled gilthead seabream GH to dolphin fish liver microsomal fraction resulted in high specific binding characterized by a Ka of 1.93 nM and a Bmax of 540 fmol/mg microsomal fraction protein. Recombinant gilthead seabream Growth Hormone, like ovine placental lactogen, exhibited growth-stimulating activity when applied orally to Sparus aurata larvae or intraperitoneally to juvenile fish.
Product Type Recombinant Protein
Cross Reactivity Gilthead Seabream
Species E. coli
Protein Tag Untagged
Sequence MDRVVLMLSV MSLGVSSQPI TDGQRLFSIA VSRVQHLHLL AQRLFSDFES SLQTEEQRQL NKIFLQDFCN SDYIISPIDK HETQRSSVLK LLSISYRLVE SWEFPSRSLS GGSAPRNQIS PKLSELKTGI HLLIRANEDG AEIFPDSSAL QLAPYGNYYQ SLGTDESLRR TYELLACFKK DMHKVETYLT VAKCRLSPEA NCTL
Buffer The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02% NaHCO3.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.