missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Guinea Pig IL-17/IL-17A Protein

Product Code. 18608177 Shop All Bio Techne Products
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 18608177

Brand: Novus Biologicals™ NBP311059

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA
Formulation PBS
Gene ID (Entrez) 3605
Molecular Weight (g/mol) MolecularWeight-theroretical: 17 kDa
Quantity 5 μg
Storage Requirements Store at -20°C.
Gene Alias CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17 Acytotoxic T-lymphocyte-associated protein 8, IL-17 CTLA-8, IL17interleukin-17 A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17 A
Gene Symbol IL17A
Conjugate Unconjugated
Species Guinea Pig
Recombinant Recombinant
Reconstitution Reconstitute with 20 uL sterile PBS containing 0.1% carrier protein. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. We recommend that the vial is gently mixed after reconstitution.)
Form Purified
Research Category Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Purity or Quality Grade >95%, by SDS-PAGE
Protein IL-17/IL-17 A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.