missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant H. pylori Fucosyltransferase Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to transfer fucose from GDP-fucose to N -Acetyllactosamine The specific activity is >1500 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | AAD07710.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Molecular Weight (g/mol) | M.W.-observed: 39 kDa, under reducing conditions., M.W.-theroretical: 46 kDa |
| Quantity | 20 μg |
| Source | E. coli-derived h. pylori Fucosyltransferase protein Met1-Tyr392, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction