missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Hepatitis B Antigen Protein

Product Code. 18647248 Shop All Bio Techne Products
Click to view available options
:
50 μg
This item is not returnable. View return policy

Product Code. 18647248

Brand: Novus Biologicals™ NBP262481

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) ELISA
Formulation PBS, 50% Glycerol
Molecular Weight (g/mol) MolecularWeight-theroretical: 18 kDa
Quantity 50 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias HBV x antigen, HBx, HBxAg
Purity or Quality Grade >95%, by SDS-PAGE
Protein Hepatitis B Antigen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.