missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis B HBV core Protein

A cDNA sequence encoding the HBV core was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12130-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939468

  • € 1815.00 / 1mg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis B HBV core Protein
Research Use Only
Recombinant Protein
E. coli
25mM Tris-HCl pH 8.0, 1.5 mM EDTA, 1.5mM Urea and 50% glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis B
Untagged
HBV Core protein is >90% pure as determined by 10% PAGE (Coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers