missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-1a Protein

A cDNA sequence encoding the HCV NS3 Genotype-1a (Amino Acids 1356-1459) was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP14026-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935449

  • € 1600.00 / 1mg

Estimated Shipment: 02-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Hepatitis C HCV NS3 Genotype-1a Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
GST
HCV NS3 Genotype-1a protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers