Learn More
enQuireBio™ Recombinant hepatitis Protein ORF3 Protein
Product Code. 16088980
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16088980
Brand: enQuireBio™ QP7492ec1mg
A DNA sequence encoding the Hepatitis E virus genotype 1 (isolate Human/Pakistan/Sar-55) (HEV-1) Protein ORF3, was expressed in the hosts and tags indicated.
Specifications
Name hepatitis Protein ORF3 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ORF3
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Hepatitis
Species E. coli
Protein Tag His-SUMO
Sequence MGSRPWALGL FCCCSSCFCL CCSRHRPVSR LAAVVGGAAA VPAVVSGVTG LILSPSQSPI FIQPTPSPRM SPLRPGLDLV FANPSDHSAP LGATRPSAPP LPHVVDLPQL GPRR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.