missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 gp120 LAV Protein

Product Code. 15909818
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
This item is not returnable. View return policy

Product Code. 15909818

Brand: enQuireBio™ QP12245100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the HIV-1 gp120 LAV was constructed and used to recombinantly synthesize the protein.

Specifications

Name HIV-1 gp120 LAV Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity HIV
Species Baculovirus
Protein Tag Untagged
Sequence IPGEKLWVTV YYGVPVWKEA TTTLFCASDA KAYDTEVHNV ATHACVPTDP NPQEVVLVNV TENFNMWKND MVEQMHEDII SLWDQSLKPC VKLTPLCVSL KCTDLKNDTN TNSSSGRMIM EKGEIKNCSF NISTSIRDKV QKEYAFFYKL DIVPIDNTSY RLISCNTSVI TQACPKVSFE PIPIHYCAPA GFAILKCNNK TFNGTGPCTN VSTVQCTHGI RPVVSTQLLL NGSLAEEDVV IRSANFTDNA KTIIVQLNTS VEINCTRPNN NTRKSIRIQR GPGRAFVTIG KIGNMRQAHC NISRAKWNAT LKQIASKLRE QFGNNKTIIF KQSSGGDPEI VTHSFNCGGE FFYCNSTQLF NSTWFNSTWS TEGSNNTEGS DTITLPCRIK QFINMWQEVG KAMYAPPISG QIRCSSNITG LLLTRDGGNN NNGSEIFRPG GGDMRDNWRS ELYKYKVVKI EPLGVAPTKA KRRVVQREKR
Buffer The protein solution contains 10mM Tris-Cl pH 7.6, 150mM NaCl and 0.01% Triton N-101.
Purity or Quality Grade Greater than 90.0% as determined by HPLC analysis and SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.