missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Horse Major allergen Equ c 1 Protein

A DNA sequence encoding the Equus caballus (Horse) Major allergen Equ c 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7892-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16071431

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001075966.1
Major allergen Equ c 1 Protein
Research Use Only
LOC100034197
Recombinant Protein
E. coli
QQEENSDVAI RNFDISKISG EWYSIFLASD VKEKIEENGS MRVFVDVIRA LDNSSLYAEY QTKVNGECTE FPMVFDKTEE DGVYSLNYDG YNVFRISEFE NDEHIILYLV NFDKDRPFQL FEFYAREPDV SPEIKEEFVK IVQKRGIVKE NIIDLTKIDR CFQLRGNGVA QA
Greater than 90% as determined by SDS-PAGE.
100034197
500 μg
Not determined.
Full Length
Horse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers