missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse Major allergen Equ c 1 Protein
A DNA sequence encoding the Equus caballus (Horse) Major allergen Equ c 1, was expressed in the hosts and tags indicated.
€ 352.00 - € 3175.00
Specifications
Accession Number | NP_001075966.1 |
---|---|
Gene ID (Entrez) | 100034197 |
Name | Major allergen Equ c 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091431
|
enQuireBio™
QP7892-YE-100UG |
100 μg |
€ 1100.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031441
|
enQuireBio™
QP7892-YE-500UG |
500 μg |
€ 2650.00
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001441
|
enQuireBio™
QP7892-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021441
|
enQuireBio™
QP7892-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041441
|
enQuireBio™
QP7892-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011441
|
enQuireBio™
QP7892-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001075966.1 | |
Major allergen Equ c 1 Protein | |
Not determined. | |
Full Length | |
Horse | |
His | |
Tris-based buffer, 50% glycerol |
100034197 | |
Research Use Only | |
LOC100034197 | |
Recombinant Protein | |
Yeast | |
QQEENSDVAI RNFDISKISG EWYSIFLASD VKEKIEENGS MRVFVDVIRA LDNSSLYAEY QTKVNGECTE FPMVFDKTEE DGVYSLNYDG YNVFRISEFE NDEHIILYLV NFDKDRPFQL FEFYAREPDV SPEIKEEFVK IVQKRGIVKE NIIDLTKIDR CFQLRGNGVA QA | |
Greater than 90% as determined by SDS-PAGE. |