missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 3-ketoacyl-CoA thiolase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 3-ketoacyl-CoA thiolase, mitochondrial, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_006102.2 |
---|---|
Gene ID (Entrez) | 10449 |
Name | 3-ketoacyl-CoA thiolase, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15916369
|
enQuireBio™
QP5603-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936369
|
enQuireBio™
QP5603-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926369
|
enQuireBio™
QP5603-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15946369
|
enQuireBio™
QP5603-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15956369
|
enQuireBio™
QP5603-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15966369
|
enQuireBio™
QP5603-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006102.2 | |
3-ketoacyl-CoA thiolase, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
10449 | |
Research Use Only | |
ACAA2 | |
Recombinant Protein | |
E. coli | |
FGAYGGLLKD FTATDLSEFA AKAALSAGKV SPETVDSVIM GNVLQSSSDA IYLARHVGLR VGIPKETPAL TINRLCGSGF QSIVNGCQEI CVKEAEVVLC GGTESMSQAP YCVRNVRFGT KLGSDIKLED SLWVSLTDQH VQLPMAMTAE NLAVKHKISR EECDKYALQS QQRWKAANDA GYFNDEMAPI EVKTKKGKQT MQVDEHARPQ TTLEQLQKLP PVFKKDGTVT AGNASGVADG AGAVIIASED AVKKHNFTPL ARIVGYFVSG CDPSIMGIGP VPAISGALKK AGLSLKDMDL VEVNEAFAPQ YLAVERSLDL DISKTNVNGG AIALGHPLGG SGSRITAHLV HELRRRGGKY AVGSACIGGG QGIAVIIQST A | |
Greater than 90% as determined by SDS-PAGE. |