missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human A2LD1 Protein

A cDNA sequence encoding the A2LD1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10921-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935578

  • € 212.00 / 10µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

A2LD1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MALVFVYGTL KRGQPNHRVL RDGAHGSAAF RARGRTLEPY PLVIAGEHNI PWLLHLPGSG RLVEGEVYAV DERMLRFLDD FESCPALYQR TVLRVQLLED RAPGAEEPPA PTAVQCFVYS RATFPPEWAQ LPHHDSYDSE GPHGLRYNPR ENR
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The A2LD1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers