missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ABCB8 Protein

Product Code. 15902768
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 15902768

Brand: enQuireBio™ QP10021iv100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens(Human) ABCB8, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001269220.1
Gene ID (Entrez) 11194
Name ABCB8 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ABCB8
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence GKTGQLHKAE GEYSDGYRSS SLLRAVAHLR SQLWAHLPRA PLAPRWSPSA WCWVGGALLG PMVLSKHPHL CLVALCEAEE APPASSTPHV VGSRFNWKLF WQFLHPHLLV LGVAVVLALG AALVNVQIPL LLGQLVEVVA KYTRDHVGSF MTESQNLSTH LLILYGVQGL LTFGYLVLLS HVGERMAVDM RRALFSSLLR QDITFFDANK TGQLVSRLTT DVQEFKSSFK LVISQGLRSC TQVAGCLVSL SMLSTRLTLL LMVATPALMG VGTLMGSGLR KLSRQCQEQI ARAMGVADEA LGNVRTVRAF AMEQREEERY GAELEACRCR AEELGRGIAL FQGLSNIAFN CMVLGTLFIG GSLVAGQQLT GGDLMSFLVA SQTVQRSMAN LSVLFGQVVR GLSAGARVFE YMALNPCIPL SGGCCVPKEQ LRGSVTFQNV CFSYPCRPGF EVLKDFTLTL PPGKIVALVG QSGGGKTTVA SLLERFYDPT AGVVMLDGRD LRTLDPSWLR GQVVGFISQE PVLFGTTIME NIRFGKLEAS DEEVYTAARE ANAHEFITSF PEGYNTVVGE RGTTLSGGQK QRLAIARALI KQPTVLILDE ATSALDAESE RVVQEALDRA SAGRTVLVIA HRLSTVRGAH CIVVMADGRV WEAGTH
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.