missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ABHD12B Protein

A cDNA sequence encoding the ABHD12B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10927-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15925598

  • € 212.00 / 20µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ABHD12B Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLGIWHT VPSCRGEDAK GKDCCWYEAA LRDGNPIIVY LHGSAEHRAA SHRLKLVKVL SDGGFHVLSV DYRGFGDSTG KPTEEGLTTD AICVYEWTKA RSGITPVCLW GHSLGTGVAT NAAKVLEEKG CPVDAIVLEA PFTNMWVASI NYPLLKIYRN IPGFLRTLMD ALRKDKIIFP NDENVKFLSS PLLILHGEDD RTVPLEYGKK LYEIARNAYR NKERVKMVIF PPGFQHNLLC KSPTLLITVR DFLSKQWS
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ABHD12B protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers