missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ABHD14B Protein

A DNA sequence encoding the Homo sapiens (Human) ABHD14B, was expressed in the hosts and tags indicated.

€ 160.00 - € 2385.00

Specifications

Accession Number NP_001139786.1
Gene ID (Entrez) 84836
Name ABHD14B Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16091551
View Documents Promotion Details
enQuireBio™
QP7914-EC-100UG
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16011561
View Documents Promotion Details
enQuireBio™
QP7914-EC-1MG
1 mg
€ 2385.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16001561
View Documents Promotion Details
enQuireBio™
QP7914-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16021561
View Documents Promotion Details
enQuireBio™
QP7914-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16031561
View Documents Promotion Details
enQuireBio™
QP7914-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16041561
View Documents Promotion Details
enQuireBio™
QP7914-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001139786.1
ABHD14B Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
84836
Research Use Only
ABHD14B
Recombinant Protein
E. coli
AASVEQREGT IQVQGQALFF REALPGSGQA RFSVLLLHGI RFSSETWQNL GTLHRLAQAG YRAVAIDLPG LGHSKEAAAP APIGELAPGS FLAAVVDALE LGPPVVISPS LSGMYSLPFL TAPGSQLPGF VPVAPICTDK INAANYASVK TPALIVYGDQ DPMGQTSFEH LKQLPNHRVL IMKGAGHPCY LDKPEEWHTG LLDFLQGLQ
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications