missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACLY / acly / ATP citrate lyase Protein

Product Code. 15996399
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15996399 1 mg 1mg
15986399 10 μg 10µg
15926409 50 μg 50µg
15976399 100 μg 100µg
15906409 200 μg 200µg
15916409 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15996399 Supplier enQuireBio™ Supplier No. QP5607ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) ACLY / acly / ATP citrate lyase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001087.2
Gene ID (Entrez) 47
Name ACLY / acly / ATP citrate lyase Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ACLY
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence KAISEQTGKE LLYKFICTTS AIQNRFKYAR VTPDTDWARL LQDHPWLLSQ NLVVKPDQLI KRRGKLGLVG VNLTLDGVKS WLKPRLGQEA TVGKATGFLK NFLIEPFVPH SQAEEFYVCI YATREGDYVL FHHEGGVDVG DVDAKAQKLL VGVDEKLNPE DIKKHLLVHA PEDKKEILAS FISGLFNFYE DLYFTYLEIN PLVVTKDGVY VLDLAAKVDA TADYICKVKW GDIEFPPPFG REAYPEEAYI ADLDAKSGAS LK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.