missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACSS1, mitochondrial Protein

Product Code. 16008981
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16008981

Brand: enQuireBio™ QP9911ye1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) ACSS1, mitochondrial, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001239604.1
Gene ID (Entrez) 84532
Name ACSS1, mitochondrial Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ACSS1
Protein Length Full. Length. of. Mature. Protein. 
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence ASGPSGSAPA VAAAAAQPGS YPALSAQAAR EPAAFWGPLA RDTLVWDTPY HTVWDCDFST GKIGWFLGGQ LNVSVNCLDQ HVRKSPESVA LIWERDEPGT EVRITYRELL ETTCRLANTL KRHGVHRGDR VAIYMPVSPL AVAAMLACAR IGAVHTVIFA GFSAESLAGR INDAKCKVVI TFNQGLRGGR VVELKKIVDE AVKHCPTVQH VLVAHRTDNK VHMGDLDVPL EQEMAKEDPV CAPESMGSED MLFMLYTSGS TGMPKGIVHT QAGYLLYAAL THKLVFDHQP GDIFGCVADI GWITGHSYVV YGPLCNGATS VLFESTPVYP NAGRYWETVE RLKINQFYGA PTAVRLLLKY GDAWVKKYDR SSLRTLGSVG EPINCEAWEW LHRVVGDSRC TLVDTWWQTE TGGICIAPRP SEEGAEILPA MAMRPFFGIV PVLMDEKGSV VEGSNVSGAL CISQAWPGMA RTIYGDHQRF VDAYFKAYPG YYFTGDGAYR TEGGYYQITG RMDDVINISG HRLGTAEIED AIADHPAVPE SAVIGYPHDI KGEAAFAFIV VKDSAGDSDV VVQELKSMVA TKIAKYAVPD EILVVKRLPK TRSGKVMRRL LRKIITSEAQ ELGDTTTLED PSIIAEILSV YQKCKDKQAA AK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 94.5%% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.