missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACSS1, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) ACSS1, mitochondrial, was expressed in the hosts and tags indicated.

€ 245.00 - € 3405.00

Specifications

Accession Number NP_001239604.1
Gene ID (Entrez) 84532
Name ACSS1, mitochondrial Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16088971
View Documents Promotion Details
enQuireBio™
QP9911-YE-100UG
100 μg
€ 770.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16008981
View Documents Promotion Details
enQuireBio™
QP9911-YE-1MG
1 mg
€ 3405.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16098971
View Documents Promotion Details
enQuireBio™
QP9911-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16018981
View Documents Promotion Details
enQuireBio™
QP9911-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16028981
View Documents Promotion Details
enQuireBio™
QP9911-YE-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16038981
View Documents Promotion Details
enQuireBio™
QP9911-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001239604.1
ACSS1, mitochondrial Protein
Not determined.
Full. Length. of. Mature. Protein.
Human
His
Tris-based buffer, 50% glycerol
84532
Research Use Only
ACSS1
Recombinant Protein
Yeast
ASGPSGSAPA VAAAAAQPGS YPALSAQAAR EPAAFWGPLA RDTLVWDTPY HTVWDCDFST GKIGWFLGGQ LNVSVNCLDQ HVRKSPESVA LIWERDEPGT EVRITYRELL ETTCRLANTL KRHGVHRGDR VAIYMPVSPL AVAAMLACAR IGAVHTVIFA GFSAESLAGR INDAKCKVVI TFNQGLRGGR VVELKKIVDE AVKHCPTVQH VLVAHRTDNK VHMGDLDVPL EQEMAKEDPV CAPESMGSED MLFMLYTSGS TGMPKGIVHT QAGYLLYAAL THKLVFDHQP GDIFGCVADI GWITGHSYVV YGPLCNGATS VLFESTPVYP NAGRYWETVE RLKINQFYGA PTAVRLLLKY GDAWVKKYDR SSLRTLGSVG EPINCEAWEW LHRVVGDSRC TLVDTWWQTE TGGICIAPRP SEEGAEILPA MAMRPFFGIV PVLMDEKGSV VEGSNVSGAL CISQAWPGMA RTIYGDHQRF VDAYFKAYPG YYFTGDGAYR TEGGYYQITG RMDDVINISG HRLGTAEIED AIADHPAVPE SAVIGYPHDI KGEAAFAFIV VKDSAGDSDV VVQELKSMVA TKIAKYAVPD EILVVKRLPK TRSGKVMRRL LRKIITSEAQ ELGDTTTLED PSIIAEILSV YQKCKDKQAA AK
Greater than 94.5%% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications