missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Active ERK2 Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The specific activity of ERK2 was determined to be 24 nmol/min/mg using a myelin basic protein (MBP) substrate and was equivalent to 639 nmol/min/mg as per radiometric assay.
Specifications
Specifications
| Accession Number | NM_002745 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied in 50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol. |
| Gene ID (Entrez) | 5594 |
| Molecular Weight (g/mol) | M.W. (Observed): 68 kDa |
| Quantity | 10 μg |
| Source | E. coli-derived human ERK2 protein |
| Storage Requirements | This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.) |
| Gene Symbol | MAPK1 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction