missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Active SHP-1 (aa 205-595) Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Measured by its ability to dephosphorylate a tyrosine residue in a peptide containing the EGFR Y992 phosphorylation site (Catalog # ES006 ). The specific activity is >40 μmol/min/mg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P29350 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, Glycerol and DTT. |
| Gene ID (Entrez) | 5777 |
| Molecular Weight (g/mol) | M.W. (Observed): 40-46 kDa, reducing conditions; M.W. (theoretical): 46 kDa |
| Name | SHP-1 |
| Quantity | 10 μg |
| Source | E. coli-derived human SHP-1 protein Ala205-Lys595, with an N-terminal Met and 6-His tag (C-terminal variants predicted) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Alias | HCP, HCPH, HPTP1C, PTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction