missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACVR2B Protein
A DNA sequence encoding the Homo sapiens (Human) ACVR2B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9768-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001097.2 | |
ACVR2B Protein | |
Research Use Only | |
ACVR2B | |
Recombinant Protein | |
Yeast | |
SGRGEAETRE CIYYNANWEL ERTNQSGLER CEGEQDKRLH CYASWRNSSG TIELVKKGCW LDDFNCYDRQ ECVATEENPQ VYFCCCEGNF CNERFTHLPE AGGPEVTYEP PPTAPTLLT | |
Greater than 90% as determined by SDS-PAGE. |
93 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |